Rat PCSK9, His Tag (PC9-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 691 (Accession P59996-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Rat PCSK9, His Tag (PC9-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 691 (Accession P59996-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Rat PCSK9, His Tag (PC9-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 691 (Accession P59996-1). 1 * 1 mg ACRBPC9-R52H4-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou