Human PCSK9 (D374Y), His Tag (PCY-H5225) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession Q8N BP7-1 (D374Y)). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human PCSK9 (D374Y), His Tag (PCY-H5225) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession Q8N BP7-1 (D374Y)). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human PCSK9 (D374Y), His Tag (PCY-H5225) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession Q8N BP7-1 (D374Y)). 1 * 1 mg ACRBPCY-H5225-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou