Human PD-1, His Tag (HPLC verified) (PD1-H5221) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accessi on NP005009.2). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human PD-1, His Tag (HPLC verified) (PD1-H5221) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accessi on NP005009.2). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human PD-1, His Tag (HPLC verified) (PD1-H5221) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accessi on NP005009.2). 1 * 100 µG ACRBPD1-H5221100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou