Mouse PD-1, His Tag (PD1-M5228) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession NP032824). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Mouse PD-1, His Tag (PD1-M5228) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession NP032824). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Mouse PD-1, His Tag (PD1-M5228) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession NP032824). 1 * 1 mg ACRBPD1-M5228-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou