Mouse PD-1, His Tag (PD1-M5228) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession NP032824). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Mouse PD-1, His Tag (PD1-M5228) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession NP032824). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Mouse PD-1, His Tag (PD1-M5228) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession NP032824). 1 * 100 µG ACRBPD1-M5228100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou