PE-Labeled Human PD-1, Fc tag, His tag (recommended for neutralizing assay) (Cat. No. PD1-HP2F2) is produced via conjugation of PE t o Human PD-1, Fc tag, His tag under optimal conditions. Human PD-1, Fc tag, His tag is expressed in HEK293 cells cont 1 * 50 Tests
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
PE-Labeled Human PD-1, Fc tag, His tag (recommended for neutralizing assay) (Cat. No. PD1-HP2F2) is produced via conjugation of PE t o Human PD-1, Fc tag, His tag under optimal conditions. Human PD-1, Fc tag, His tag is expressed in HEK293 cells cont 1 * 50 Tests ACRBPD1HP2F250TEST 1 * 50 Tests
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou