Human PD-L2, His Tag (SPR verified) (PD2-H5220) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accessi on AAI13679). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human PD-L2, His Tag (SPR verified) (PD2-H5220) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accessi on AAI13679). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human PD-L2, His Tag (SPR verified) (PD2-H5220) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accessi on AAI13679). 1 * 1 mg ACRBPD2-H5220-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou