Recombinat Protein,Human PH20 SPAM1 Protein, His Tag,Human PH20, His Tag (PH0-H5225) is expressed from human 293 cells (HEK293). It contains AA Leu 36 - Tyr 482 (Accession AAH26163).This protein carries a polyhistidine tag at the C-terminus. The pr l 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human PH20 SPAM1 Protein, His Tag,Human PH20, His Tag (PH0-H5225) is expressed from human 293 cells (HEK293). It contains AA Leu 36 - Tyr 482 (Accession AAH26163).This protein carries a polyhistidine tag at the C-terminus. The pr l 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human PH20 SPAM1 Protein, His Tag,Human PH20, His Tag (PH0-H5225) is expressed from human 293 cells (HEK293). It contains AA Leu 36 - Tyr 482 (Accession AAH26163).This protein carries a polyhistidine tag at the C-terminus. The pr l 1 * 25 µG ACRBPH0-H5225-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou