Recombinat Protein,Human pIgR Protein, His Tag,Human pIgR, His Tag (PIR-H5222) is expressed from human 293 cells (HEK293). It contai ns AA Lys 19 - Arg 638 (Accession AAI10495.1).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human pIgR Protein, His Tag,Human pIgR, His Tag (PIR-H5222) is expressed from human 293 cells (HEK293). It contai ns AA Lys 19 - Arg 638 (Accession AAI10495.1).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg ACRBPIR-H5222-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou