Human PLAU, His Tag (PLU-H5229) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Leu 431 (Accession NP002649.1). 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Human PLAU, His Tag (PLU-H5229) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Leu 431 (Accession NP002649.1). 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Human PLAU, His Tag (PLU-H5229) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Leu 431 (Accession NP002649.1). 1 * 50 µG ACRBPLU-H5229-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou