Recombinat Protein,Human PSGL-1 CD162 Protein, His Tag,Human PSGL-1, His Tag (PS1-H52H9) is expressed from human 293 cells (HEK293) . It contains AA Gln 42 - Cys 320 (Accession Q14242-1).This protein carries a polyhistidine tag at the C-terminus. Th 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human PSGL-1 CD162 Protein, His Tag,Human PSGL-1, His Tag (PS1-H52H9) is expressed from human 293 cells (HEK293) . It contains AA Gln 42 - Cys 320 (Accession Q14242-1).This protein carries a polyhistidine tag at the C-terminus. Th 1 * 1 mg ACRBPS1-H52H9-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou