Recombinat Protein,Human PSME3 PA28g Protein, His Tag,Human PSME3, His Tag (PS3-H5142) is expressed from E.coli cells. It contains AA Met 1 - Tyr 254 (Accession NP00578).This protein carries a polyhistidine tag at the N-terminus. The protein has a c 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human PSME3 PA28g Protein, His Tag,Human PSME3, His Tag (PS3-H5142) is expressed from E.coli cells. It contains AA Met 1 - Tyr 254 (Accession NP00578).This protein carries a polyhistidine tag at the N-terminus. The protein has a c 1 * 1 mg ACRBPS3-H5142-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou