Recombinat Protein,Human PSCA Protein, His Tag,Human PSCA, His Tag (PSA-H52H6) is expressed from human 293 cells (HEK293). It contai ns AA Leu 12 - Ser 86 (Accession O43653-1).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human PSCA Protein, His Tag,Human PSCA, His Tag (PSA-H52H6) is expressed from human 293 cells (HEK293). It contai ns AA Leu 12 - Ser 86 (Accession O43653-1).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 100 µG ACRBPSA-H52H6100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou