Recombinat Protein,FITC-Labeled Human PSMA FOLH1 Protein, His Tag,FITC-Labeled Human PSMA, His Tag (PSA-HF244) is expressed from hu man 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession Q04609-1). It is the FITC labeled form of Human PS 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,FITC-Labeled Human PSMA FOLH1 Protein, His Tag,FITC-Labeled Human PSMA, His Tag (PSA-HF244) is expressed from hu man 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession Q04609-1). It is the FITC labeled form of Human PS 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,FITC-Labeled Human PSMA FOLH1 Protein, His Tag,FITC-Labeled Human PSMA, His Tag (PSA-HF244) is expressed from hu man 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession Q04609-1). It is the FITC labeled form of Human PS 1 * 200 µG ACRBPSA-HF244200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou