Recombinat Protein,Mouse PSMA FOLH1 Protein, His Tag,Mouse PSMA, His Tag (PSA-M5245) is expressed from human 293 cells (HEK293). It contains AA Ile 44 - Ala 752 (Accession O35409-1).This protein carries a polyhistidine tag at the N-terminus. The pr l 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Mouse PSMA FOLH1 Protein, His Tag,Mouse PSMA, His Tag (PSA-M5245) is expressed from human 293 cells (HEK293). It contains AA Ile 44 - Ala 752 (Accession O35409-1).This protein carries a polyhistidine tag at the N-terminus. The pr l 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse PSMA FOLH1 Protein, His Tag,Mouse PSMA, His Tag (PSA-M5245) is expressed from human 293 cells (HEK293). It contains AA Ile 44 - Ala 752 (Accession O35409-1).This protein carries a polyhistidine tag at the N-terminus. The pr l 1 * 1 mg ACRBPSA-M5245-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou