Recombinat Protein,Human PTPRD Protein, His Tag,Human PTPRD, His Tag (PTD-H52H9) is expressed from human 293 cells (HEK293). It cont ains AA Glu 21 - Glu 1265 (Accession P23468-1).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human PTPRD Protein, His Tag,Human PTPRD, His Tag (PTD-H52H9) is expressed from human 293 cells (HEK293). It cont ains AA Glu 21 - Glu 1265 (Accession P23468-1).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human PTPRD Protein, His Tag,Human PTPRD, His Tag (PTD-H52H9) is expressed from human 293 cells (HEK293). It cont ains AA Glu 21 - Glu 1265 (Accession P23468-1).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 1 mg ACRBPTD-H52H9-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou