Recombinat Protein,Human Reg4 Protein, His Tag,Human Reg4, His Tag (RE4-H5221) is expressed from human 293 cells (HEK293). It contai ns AA Asp 23 - Pro 158 (Accession AAH17089).This protein carries a polyhistidine tag at the C-terminus. The protein e 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Reg4 Protein, His Tag,Human Reg4, His Tag (RE4-H5221) is expressed from human 293 cells (HEK293). It contai ns AA Asp 23 - Pro 158 (Accession AAH17089).This protein carries a polyhistidine tag at the C-terminus. The protein e 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Reg4 Protein, His Tag,Human Reg4, His Tag (RE4-H5221) is expressed from human 293 cells (HEK293). It contai ns AA Asp 23 - Pro 158 (Accession AAH17089).This protein carries a polyhistidine tag at the C-terminus. The protein e 1 * 1 mg ACRBRE4-H5221-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou