Recombinat Protein,Human RENIN Protein, His Tag,Human RENIN, His Tag (REN-H5221) is expressed from human 293 cells (HEK293). It cont ains AA Leu 24 - Arg 406 (Accession AAH33474).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human RENIN Protein, His Tag,Human RENIN, His Tag (REN-H5221) is expressed from human 293 cells (HEK293). It cont ains AA Leu 24 - Arg 406 (Accession AAH33474).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 100 µG ACRBREN-H5221100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou