Recombinat Protein,Mouse RENIN Protein, His Tag,Mouse RENIN, His Tag (REN-M5222) is expressed from human 293 cells (HEK293). It cont ains AA Leu 22 - Arg 402 (Accession NP112469.1).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse RENIN Protein, His Tag,Mouse RENIN, His Tag (REN-M5222) is expressed from human 293 cells (HEK293). It cont ains AA Leu 22 - Arg 402 (Accession NP112469.1).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg ACRBREN-M5222-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou