Human ROR1 (39-151, Ig-like domain), His Tag (RO1-H5221) is expressed from human 293 cells (HEK293). It contains AA Glu 39 - Gly 151 (Accession Q01973-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human ROR1 (39-151, Ig-like domain), His Tag (RO1-H5221) is expressed from human 293 cells (HEK293). It contains AA Glu 39 - Gly 151 (Accession Q01973-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human ROR1 (39-151, Ig-like domain), His Tag (RO1-H5221) is expressed from human 293 cells (HEK293). It contains AA Glu 39 - Gly 151 (Accession Q01973-1). 1 * 1 mg ACRBRO1-H5221-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou