Human ROR1, His Tag (RO1-H522y) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession Q01973-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human ROR1, His Tag (RO1-H522y) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession Q01973-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human ROR1, His Tag (RO1-H522y) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession Q01973-1). 1 * 1 mg ACRBRO1-H522Y-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou