Recombinat Protein,Rat ROR1 Protein, His Tag,Rat ROR1, His Tag (RO1-R5221) is expressed from human 293 cells (HEK293). It contains A A Gln 30 - Glu 403 (Accession EDL97819).This protein carries a polyhistidine tag at the C-terminus. The protein has 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Rat ROR1 Protein, His Tag,Rat ROR1, His Tag (RO1-R5221) is expressed from human 293 cells (HEK293). It contains A A Gln 30 - Glu 403 (Accession EDL97819).This protein carries a polyhistidine tag at the C-terminus. The protein has 1 * 1 mg ACRBRO1-R5221-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou