Recombinat Protein,Biotinylated Human ROR2 NTRKR2 Protein, His,Avitag™,Biotinylated Human ROR2, His,Avitag (RO2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession A1L4F5-1).This protein carries a polyhisti e 1 * 25 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Biotinylated Human ROR2 NTRKR2 Protein, His,Avitag™,Biotinylated Human ROR2, His,Avitag (RO2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession A1L4F5-1).This protein carries a polyhisti e 1 * 25 µG ACRBRO2-H82E3-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou