Recombinat Protein,Biotinylated Human ROR2 NTRKR2 Protein, His,Avitag,Biotinylated Human ROR2, His,Avitag (RO2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession A1L4F5-1).This protein carries a polyhisti e 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human ROR2 NTRKR2 Protein, His,Avitag,Biotinylated Human ROR2, His,Avitag (RO2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession A1L4F5-1).This protein carries a polyhisti e 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Biotinylated Human ROR2 NTRKR2 Protein, His,Avitag™,Biotinylated Human ROR2, His,Avitag (RO2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession A1L4F5-1).This protein carries a polyhisti e 1 * 200 µG ACRBRO2-H82E3200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou