Recombinant Protein G, His Tag, also known as Recombinant Staphylococcus Aureus protein G148, was expressed in E.Coli at ACRObiosyst ems. 1 * 10 mg
Fournisseur: ACROBIOSYSTEMS
Recombinant Protein G, His Tag, also known as Recombinant Staphylococcus Aureus protein G148, was expressed in E.Coli at ACRObiosyst ems. 1 * 10 mg
COMMANDE
Développer tout / Réduire tout
Recombinant Protein G, His Tag, also known as Recombinant Staphylococcus Aureus protein G148, was expressed in E.Coli at ACRObiosyst ems. 1 * 10 mg ACRBRPG-S3140-10MG 1 * 10 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou