Recombinat Protein,Recombinant Protein L Protein, His Tag,Recombinant Protein L, His Tag, also known as Recombinant Peptostreptococc us magnus Protein L, was expressed in E.Coli at ACRObiosystems. ,Recombinant Protein L, His Tag is fused with the pol 1 * 10 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Recombinant Protein L Protein, His Tag,Recombinant Protein L, His Tag, also known as Recombinant Peptostreptococc us magnus Protein L, was expressed in E.Coli at ACRObiosystems. ,Recombinant Protein L, His Tag is fused with the pol 1 * 10 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Recombinant Protein L Protein, His Tag,Recombinant Protein L, His Tag, also known as Recombinant Peptostreptococc us magnus Protein L, was expressed in E.Coli at ACRObiosystems. ,Recombinant Protein L, His Tag is fused with the pol 1 * 10 mg ACRBRPL-P3141-10MG 1 * 10 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou