Recombinat Protein,Human R-Spondin 3 RSPO3 (22-146) Protein, His Tag,Human R-Spondin 3 (22-146), His Tag (RS6-H5222) is expressed f rom human 293 cells (HEK293). It contains AA Gln 22 - Val 146 (Accession AAH22367).This protein carries a polyhistidi 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human R-Spondin 3 RSPO3 (22-146) Protein, His Tag,Human R-Spondin 3 (22-146), His Tag (RS6-H5222) is expressed f rom human 293 cells (HEK293). It contains AA Gln 22 - Val 146 (Accession AAH22367).This protein carries a polyhistidi 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human R-Spondin 3 RSPO3 (22-146) Protein, His Tag,Human R-Spondin 3 (22-146), His Tag (RS6-H5222) is expressed f rom human 293 cells (HEK293). It contains AA Gln 22 - Val 146 (Accession AAH22367).This protein carries a polyhistidi 1 * 50 µG ACRBRS6-H5222-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou