Recombinat Protein,HRSV (A) Glycoprotein G Protein, His Tag,HRSV (A) Glycoprotein G, His Tag (RSG-V5221) is expressed from human 293 cells (HEK293). It contains AA His 67 - Arg 297 (Accession P20895).This protein carries a polyhistidine tag at the p 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,HRSV (A) Glycoprotein G Protein, His Tag,HRSV (A) Glycoprotein G, His Tag (RSG-V5221) is expressed from human 293 cells (HEK293). It contains AA His 67 - Arg 297 (Accession P20895).This protein carries a polyhistidine tag at the p 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,HRSV (A) Glycoprotein G Protein, His Tag,HRSV (A) Glycoprotein G, His Tag (RSG-V5221) is expressed from human 293 cells (HEK293). It contains AA His 67 - Arg 297 (Accession P20895).This protein carries a polyhistidine tag at the p 1 * 50 µG ACRBRSG-V5221-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou