Recombinat Protein,Human S100A10 Protein, His Tag,Human S100A10, His Tag (S10-H5127) is expressed from E.coli cells. It contains AA Met 1 - Lys 97 (Accession AAH15973).This protein carries a polyhistidine tag at the C-terminus. The protein has a cal 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human S100A10 Protein, His Tag,Human S100A10, His Tag (S10-H5127) is expressed from E.coli cells. It contains AA Met 1 - Lys 97 (Accession AAH15973).This protein carries a polyhistidine tag at the C-terminus. The protein has a cal 1 * 100 µG ACRBS10-H5127100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou