Recombinat Protein,Human S100A6 Protein, Tag Free,Human S100A6, Tag Free (S16-H5111) is expressed from E.coli cells. It contains AA Met 1 - Gly 90 (Accession AAH01431).This protein carries no tag. The protein has a calculated MW of 10.2 kDa. The pro 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human S100A6 Protein, Tag Free,Human S100A6, Tag Free (S16-H5111) is expressed from E.coli cells. It contains AA Met 1 - Gly 90 (Accession AAH01431).This protein carries no tag. The protein has a calculated MW of 10.2 kDa. The pro 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human S100A6 Protein, Tag Free,Human S100A6, Tag Free (S16-H5111) is expressed from E.coli cells. It contains AA Met 1 - Gly 90 (Accession AAH01431).This protein carries no tag. The protein has a calculated MW of 10.2 kDa. The pro 1 * 1 mg ACRBS16-H5111-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou