Recombinat Protein,Human S100A8 Protein, Tag Free,Human S100A8, Tag Free (S18-H5112) is expressed from E.coli cells. It contains AA Met 1 - Glu 93 (Accession AAH05928).This protein carries no tag. The protein has a calculated MW of 10.8 kDa. The pro 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human S100A8 Protein, Tag Free,Human S100A8, Tag Free (S18-H5112) is expressed from E.coli cells. It contains AA Met 1 - Glu 93 (Accession AAH05928).This protein carries no tag. The protein has a calculated MW of 10.8 kDa. The pro 1 * 1 mg ACRBS18-H5112-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou