Recombinat Protein,Human S100B Protein, His Tag,Human S100B, His Tag (S1B-H5143) is expressed from E.coli cells. It contains AA Met 1 - Glu 92 (Accession NP006263).This protein carries a polyhistidine tag at the N-terminus. The protein has a calcula 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human S100B Protein, His Tag,Human S100B, His Tag (S1B-H5143) is expressed from E.coli cells. It contains AA Met 1 - Glu 92 (Accession NP006263).This protein carries a polyhistidine tag at the N-terminus. The protein has a calcula 1 * 1 mg ACRBS1B-H5143-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou