Recombinat Protein,Human S100P S100E Protein, Tag Free,Human S100P, Tag Free (S1P-H5110) is expressed from E.coli cells. It contain s AA Met 1 - Lys 95 (Accession AAH06819).This protein carries no tag. The protein has a calculated MW of 10.4 kDa. gra 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human S100P S100E Protein, Tag Free,Human S100P, Tag Free (S1P-H5110) is expressed from E.coli cells. It contain s AA Met 1 - Lys 95 (Accession AAH06819).This protein carries no tag. The protein has a calculated MW of 10.4 kDa. gra 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human S100P S100E Protein, Tag Free,Human S100P, Tag Free (S1P-H5110) is expressed from E.coli cells. It contain s AA Met 1 - Lys 95 (Accession AAH06819).This protein carries no tag. The protein has a calculated MW of 10.4 kDa. gra 1 * 1 mg ACRBS1P-H5110-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou