Recombinat Protein,Human SCARB1 SR-B1 Protein, Fc Tag,Human SCARB1, Fc Tag (SC1-H5258) is expressed from human 293 cells (HEK293). It contains AA Pro 33 - Tyr 443 (Accession NP005496.4).This protein carries a human IgG1 Fc tag at the C-terminus. The 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human SCARB1 SR-B1 Protein, Fc Tag,Human SCARB1, Fc Tag (SC1-H5258) is expressed from human 293 cells (HEK293). It contains AA Pro 33 - Tyr 443 (Accession NP005496.4).This protein carries a human IgG1 Fc tag at the C-terminus. The 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human SCARB1 SR-B1 Protein, Fc Tag,Human SCARB1, Fc Tag (SC1-H5258) is expressed from human 293 cells (HEK293). It contains AA Pro 33 - Tyr 443 (Accession NP005496.4).This protein carries a human IgG1 Fc tag at the C-terminus. The 1 * 1 mg ACRBSC1-H5258-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou