Recombinat Protein,Human SCARB2 SR-B2 LIMPII Protein, Fc Tag,Human SCARB2, Fc Tag (SC2-H5250) is expressed from human 293 cells (H EK293). It contains AA Arg 27 - Thr 432 (Accession AAH21892).This protein carries a human IgG1 Fc tag at the C-terminu 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human SCARB2 SR-B2 LIMPII Protein, Fc Tag,Human SCARB2, Fc Tag (SC2-H5250) is expressed from human 293 cells (H EK293). It contains AA Arg 27 - Thr 432 (Accession AAH21892).This protein carries a human IgG1 Fc tag at the C-terminu 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human SCARB2 SR-B2 LIMPII Protein, Fc Tag,Human SCARB2, Fc Tag (SC2-H5250) is expressed from human 293 cells (H EK293). It contains AA Arg 27 - Thr 432 (Accession AAH21892).This protein carries a human IgG1 Fc tag at the C-terminu 1 * 100 µG ACRBSC2-H5250100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou