Mouse SCF, His Tag (SCF-M5228) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Ala 189 (Accession NP038626). 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Mouse SCF, His Tag (SCF-M5228) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Ala 189 (Accession NP038626). 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Mouse SCF, His Tag (SCF-M5228) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Ala 189 (Accession NP038626). 1 * 50 µG ACRBSCF-M5228-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou