Recombinat Protein,Human Serpin F1 PEDF Protein, His Tag,Human Serpin F1, His Tag (SE1-H5221) is expressed from human 293 cells (HE K293). It contains AA Gln 20 - Pro 418 (Accession NP002606).This protein carries a polyhistidine tag at the C-terminu 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Serpin F1 PEDF Protein, His Tag,Human Serpin F1, His Tag (SE1-H5221) is expressed from human 293 cells (HE K293). It contains AA Gln 20 - Pro 418 (Accession NP002606).This protein carries a polyhistidine tag at the C-terminu 1 * 100 µG ACRBSE1-H5221100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou