Recombinat Protein,Human Serpin A1 Protein, His Tag,Human Serpin A1, His Tag (SE1-H5226) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Lys 418 (Accession AAH11991).This protein carries a polyhistidine tag at the C-terminus. Th 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Serpin A1 Protein, His Tag,Human Serpin A1, His Tag (SE1-H5226) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Lys 418 (Accession AAH11991).This protein carries a polyhistidine tag at the C-terminus. Th 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Serpin A1 Protein, His Tag,Human Serpin A1, His Tag (SE1-H5226) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Lys 418 (Accession AAH11991).This protein carries a polyhistidine tag at the C-terminus. Th 1 * 50 µG ACRBSE1-H5226-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou