Recombinat Protein,Human Serpin E2 PN1 Protein, His Tag,Human Serpin E2, His Tag (SE2-H5225) is expressed from human 293 cells (HEK 293). It contains AA Ser 20 - Pro 397 (Accession P07093-2).This protein carries a polyhistidine tag at the C-terminus 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Serpin E2 PN1 Protein, His Tag,Human Serpin E2, His Tag (SE2-H5225) is expressed from human 293 cells (HEK 293). It contains AA Ser 20 - Pro 397 (Accession P07093-2).This protein carries a polyhistidine tag at the C-terminus 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Serpin E2 PN1 Protein, His Tag,Human Serpin E2, His Tag (SE2-H5225) is expressed from human 293 cells (HEK 293). It contains AA Ser 20 - Pro 397 (Accession P07093-2).This protein carries a polyhistidine tag at the C-terminus 1 * 25 µG ACRBSE2-H5225-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou