Recombinat Protein,Human Semaphorin 4A SEMA4A Protein, His Tag,Human Semaphorin 4A, His Tag (SEA-H5224) is expressed from human 293 cells (HEK293). It contains AA Gly 33 - His 683 (Accession AAH20974).This protein carries a polyhistidine tag at the h 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Semaphorin 4A SEMA4A Protein, His Tag,Human Semaphorin 4A, His Tag (SEA-H5224) is expressed from human 293 cells (HEK293). It contains AA Gly 33 - His 683 (Accession AAH20974).This protein carries a polyhistidine tag at the h 1 * 100 µG ACRBSEA-H5224100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou