Recombinat Protein,Human P-Selectin CD62P Protein, Fc Tag,Human P-Selectin, Fc Tag (SEP-H5255) is expressed from human 293 cells (H EK293). It contains AA Trp 42 - Ala 771 (Accession AAN06828).This protein carries a human IgG1 Fc tag at the C-termin 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human P-Selectin CD62P Protein, Fc Tag,Human P-Selectin, Fc Tag (SEP-H5255) is expressed from human 293 cells (H EK293). It contains AA Trp 42 - Ala 771 (Accession AAN06828).This protein carries a human IgG1 Fc tag at the C-termin 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human P-Selectin CD62P Protein, Fc Tag,Human P-Selectin, Fc Tag (SEP-H5255) is expressed from human 293 cells (H EK293). It contains AA Trp 42 - Ala 771 (Accession AAN06828).This protein carries a human IgG1 Fc tag at the C-termin 1 * 1 mg ACRBSEP-H5255-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou