Recombinat Protein,Human P-Selectin CD62P Protein, His Tag,Human P-Selectin, His Tag (SEP-H52E4) is expressed from human 293 cells (HEK293). It contains AA Trp 42 - Ala 771 (Accession AAN06828).This protein carries a polyhistidine tag at the C-termi 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human P-Selectin CD62P Protein, His Tag,Human P-Selectin, His Tag (SEP-H52E4) is expressed from human 293 cells (HEK293). It contains AA Trp 42 - Ala 771 (Accession AAN06828).This protein carries a polyhistidine tag at the C-termi 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human P-Selectin CD62P Protein, His Tag,Human P-Selectin, His Tag (SEP-H52E4) is expressed from human 293 cells (HEK293). It contains AA Trp 42 - Ala 771 (Accession AAN06828).This protein carries a polyhistidine tag at the C-termi 1 * 100 µG ACRBSEP-H52E4100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou