FITC-Labeled Human Siglec-2, His Tag (Cat. No. SI2-HF2H6) is expressed from human HEK293 cells. It contains AA Asp 20 - Arg 687 (Acc ession P20273-1). It is the FITC labeled form of Human Siglec-2, His Tag (Cat. No. CD2-H52H8). 1 * 200 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
FITC-Labeled Human Siglec-2, His Tag (Cat. No. SI2-HF2H6) is expressed from human HEK293 cells. It contains AA Asp 20 - Arg 687 (Acc ession P20273-1). It is the FITC labeled form of Human Siglec-2, His Tag (Cat. No. CD2-H52H8). 1 * 200 µG ACRBSI2-HF2H6200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou