Recombinat Protein,Rhesus macaque Siglec-2 CD22 Protein, His Tag,Rhesus macaque Siglec-2, His Tag (SI2-R52Ha) is expressed from hum an 293 cells (HEK293). It contains AA Ser 20 - Arg 686 (Accession EHH29920.1).This protein carries a polyhistidine ta 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Rhesus macaque Siglec-2 CD22 Protein, His Tag,Rhesus macaque Siglec-2, His Tag (SI2-R52Ha) is expressed from hum an 293 cells (HEK293). It contains AA Ser 20 - Arg 686 (Accession EHH29920.1).This protein carries a polyhistidine ta 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Rhesus macaque Siglec-2 CD22 Protein, His Tag,Rhesus macaque Siglec-2, His Tag (SI2-R52Ha) is expressed from hum an 293 cells (HEK293). It contains AA Ser 20 - Arg 686 (Accession EHH29920.1).This protein carries a polyhistidine ta 1 * 1 mg ACRBSI2-R52HA-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou