Recombinat Protein,Human Siglec-6 CD327 Protein, Fc Tag,Human Siglec-6, Fc Tag (SI6-H5256) is expressed from human 293 cells (HEK29 3). It contains AA Gln 16 - Val 320 (Accession AAH35359).This protein carries a human IgG1 Fc tag at the C-terminus. s 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Siglec-6 CD327 Protein, Fc Tag,Human Siglec-6, Fc Tag (SI6-H5256) is expressed from human 293 cells (HEK29 3). It contains AA Gln 16 - Val 320 (Accession AAH35359).This protein carries a human IgG1 Fc tag at the C-terminus. s 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Siglec-6 CD327 Protein, Fc Tag,Human Siglec-6, Fc Tag (SI6-H5256) is expressed from human 293 cells (HEK29 3). It contains AA Gln 16 - Val 320 (Accession AAH35359).This protein carries a human IgG1 Fc tag at the C-terminus. s 1 * 100 µG ACRBSI6-H5256100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou