Recombinat Protein,Cynomolgus Siglec-9 Protein, His Tag,Cynomolgus Siglec-9, His Tag (SI9-C52H9) is expressed from human 293 cells ( HEK293). It contains AA Gln 20 - Gly 350 (Accession XP005590140.1).This protein carries a polyhistidine tag at the p 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Cynomolgus Siglec-9 Protein, His Tag,Cynomolgus Siglec-9, His Tag (SI9-C52H9) is expressed from human 293 cells ( HEK293). It contains AA Gln 20 - Gly 350 (Accession XP005590140.1).This protein carries a polyhistidine tag at the p 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Cynomolgus Siglec-9 Protein, His Tag,Cynomolgus Siglec-9, His Tag (SI9-C52H9) is expressed from human 293 cells ( HEK293). It contains AA Gln 20 - Gly 350 (Accession XP005590140.1).This protein carries a polyhistidine tag at the p 1 * 1 mg ACRBSI9-C52H9-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou