Recombinat Protein,Cynomolgus Siglec-9 Protein, His Tag,Cynomolgus Siglec-9, His Tag (SI9-C52H9) is expressed from human 293 cells ( HEK293). It contains AA Gln 20 - Gly 350 (Accession XP005590140.1).This protein carries a polyhistidine tag at the p 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Cynomolgus Siglec-9 Protein, His Tag,Cynomolgus Siglec-9, His Tag (SI9-C52H9) is expressed from human 293 cells ( HEK293). It contains AA Gln 20 - Gly 350 (Accession XP005590140.1).This protein carries a polyhistidine tag at the p 1 * 100 µG ACRBSI9-C52H9100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou