Cynomolgus SIRP alpha, His Tag (SIA-C52H7) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Asn 370 (Accession N P001271679.1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Cynomolgus SIRP alpha, His Tag (SIA-C52H7) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Asn 370 (Accession N P001271679.1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Cynomolgus SIRP alpha, His Tag (SIA-C52H7) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Asn 370 (Accession N P001271679.1). 1 * 100 µG ACRBSIA-C52H7100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou