Human SIRP alpha, His Tag (HPLC-verified) (SIA-H5225) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (A ccession NP001035111). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human SIRP alpha, His Tag (HPLC-verified) (SIA-H5225) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (A ccession NP001035111). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human SIRP alpha, His Tag (HPLC-verified) (SIA-H5225) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (A ccession NP001035111). 1 * 1 mg ACRBSIA-H5225-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou