Biotinylated Human SIRP alpha, His,Avitag (HPLC-verified) (SIA-H82E0) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (Accession NP001035111). 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Biotinylated Human SIRP alpha, His,Avitag (HPLC-verified) (SIA-H82E0) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (Accession NP001035111). 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Biotinylated Human SIRP alpha, His,Avitag (HPLC-verified) (SIA-H82E0) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (Accession NP001035111). 1 * 200 µG ACRBSIA-H82E0200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou